missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC2 Mouse anti-Human, Clone: OTI2E1, lyophilized, TrueMAB™
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Myosin binding protein C (MYBPC) is a component of the thick filament of striated muscle, with the fast-type isoform designated MYBPC2. Both the fast-type and slow-type MYBPC protein contains seven immunoglobulin C2 motifs and three fibronectin type-III repeats. MYBPC2 is typically required for strong contractions and functions under anaerobic conditions. It is more similar to the cardiac isoform (MYBPC3) than to the slow-type isoform (MYBPC1) in terms of pCa50-indexed force development, length-independent cooperativity and length dependent activation. It has been suggested that in cardiac and fast muscle MYBPC2 contributes to an internal load, possibly by binding to actin via its N-terminal region.
Especificaciones
Especificaciones
| Antígeno | MYBPC2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI2E1 |
| Conjugado | Unconjugated |
| Formulación | PBS with 8% trehalose and no preservative; pH 7.3 |
| génica | MYBPC2 |
| N.º de referencia del gen | Q14324 |
| Alias de gen | MYBPC, MYBPCF |
| Símbolos de los genes | MYBPC2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?