missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Antibody (2F11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-22615
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human samples. It is validated for Western Blot, ELISA.
Especificaciones
| Muscle Phosphofructokinase/PFKM/PFK-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 2-5 ug per 10 6 cells, ELISA 1:100-1:2000 | |
| 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Anti-human PFKM mAb, clone 2F11, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PFKM protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA | |
| 2F11 | |
| Unconjugated | |
| NP_000280 | |
| PFKM | |
| Recombinant human PFKM (1-780aa) purified from E. coli | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5213 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido