missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Muromonab Chimeric Recombinant Rabbit Monoclonal Antibody (OKT-3 (muromonab))

Descripción
Specificity: Epitope on the epsilon-subunit within the human CD3 complex.
Murine monoclonal antibody specific to CD3 T-cell lymphocyte antigens. More specifically it is a purified murine (mouse) monoclonal antibody, directed against the CD3 (T3) receptor on the surface of human T-cells (T-lymphocytes) cultured using the murine ascites method. Muromonab is 93% monomeric immune globulin G type 2a (IgG2a). Muromonab binds to the T-cell surface glycoprotein CD3 epsilon chain. It appears to kill CD-3 positive cells by inducing Fc mediated apoptosis, antibody mediated cytotoxicity and complement-dependent cytotoxicity.
Especificaciones
Especificaciones
| Antígeno | Muromonab Chimeric |
| Aplicaciones | Flow Cytometry, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | OKT-3 (muromonab) |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% ProClin 300 |
| génica | Cd247 |
| N.º de referencia del gen | P04235, P11942, P22646, P24161 |
| Especie del huésped | Rabbit |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?