Descripción
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.
Especificaciones
Especificaciones
| Antígeno | MUC2 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.43 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | MUC2 |
| N.º de referencia del gen | Q02817, Q80Z19 |
| Alias de gen | Intestinal mucin 2, MLP, MUC 2, MUC2, Mucin 2, SMUC |
| Símbolos de los genes | Muc2 |
| Mostrar más |