Learn More
MTM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. p-7206245 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16852354 20 μL 20 microlitros
16842354 150 μL 150 microlitros
2 options
Código de producto. 16852354
Marca: Proteintech 139241AP20UL
Rabbit Polyclonal Antibody
Human MTM1, a 603 amino-acid protein, is mutated in myotubular myopathy. The largely related protein hMTMR2 is found mutated in a recessive form of Charcot-Marie-tooth neuropathy. Myotubularin is primarily a lipid phosphatase that acts on hosphatidylinositol 3-monophosphate and is involved in the regulation of the phosphatidylinositol 3-kinase (PI 3-kinase) pathway and membrane trafficking. Wildtype myotubularin can directly dephosphorylate PI 3-P and PI 4-P in vitro. Thus, it decreases PI 3-P levels by down-regulating PI 3-K activity and by facilitating the degradation of PI 3-P.
Especificaciones
Antígeno MTM1
Aplicaciones Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Concentración 0.35 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica MTM1
N.º de referencia del gen Q13496, Q6AXQ4, Q9Z2C5
Alias de gen CG2, CNM, MTM1, MTMX, Myotubularin, myotubularin 1, XLMTM
Símbolos de los genes Mtm1
Especie del huésped Rabbit
Inmunógeno MTM1 Fusion Protein Ag4940
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17772, 288762, 4534
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG