missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTIF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA566392
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: ELLAYDVVCED YGGDVQAVPV SALTGDNLMA LAEATVALAE MLELKADPNG PVEGTVIESF TDKGRGLVTT AIIQRGTLRK GSVLVAGKCW AKV Highest antigen sequence identity to the following orthologs - mouse 91%, rat 90%.
One of the essential components for the initiation of protein synthesis. Protects formylmethionyl-tRNA from spontaneous hydrolysis and promotes its binding to the 30S ribosomal subunits. Also involved in the hydrolysis of GTP during the formation of the 70S ribosomal complex.
Especificaciones
| MTIF2 | |
| Polyclonal | |
| Unconjugated | |
| MTIF2 | |
| IF2(mt); IF-2(Mt); IF-2mt; mitochondrial translational initiation factor 2; MTIF2; Translation initiation factor IF-2, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P46199 | |
| MTIF2 | |
| Recombinant Human MTIF2. Recombinant protein control fragment (Product #RP-107065). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido