missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFD1 Antibody, CoraFluor™ 1, Novus Biologicals™
Descripción
MTHFD1 Polyclonal antibody specifically detects MTHFD1 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | MTHFD1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | C1-THF synthase, EC 6.3.4.3, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methylenetetrahydrofolatecyclohydrolase, 10-formyltetrahydrofolate synthetase, methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, MTHFCC-1-tetrahydrofolate synthase, cytoplasmic, MTHFDcytoplasmic C-1-tetrahydrofolate synthase |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MTHFD1. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?