Learn More
Invitrogen™ MTF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA596571
Descripción
Immunogen sequence: MRDSTGAGNS LVHKRSPLRR NQKTPTSLTK LSLQDGHKAK KPACKFEEGQ DVLARWSDGL FYLGTIKKIN ILKQSCFIIF EDSSKSWVLW KDIQTGATGS GEMVCTICQE EYSEAPNEMV ICDKCGQGYH QLCHTPHIDS SVIDSDEKWL CRQCVFATTT KRGGALKKGP NAKALQVMKQ TLPYSVADLE; Positive Samples: HL-60, HepG2, Mouse brain, Rat brain; Cellular Location: Nucleus.
Polycomb group (PcG) that specifically binds histone H3 trimethylated at 'Lys-36' (H3K36me3) and recruits the PRC2 complex. Acts by binding to H3K36me3, a mark for transcriptional activation, and recruiting the PRC2 complex, leading to enhance PRC2 H3K27me3 methylation activity. Regulates the transcriptional networks during embryonic stem cell self-renewal and differentiation. Promotes recruitment of the PRC2 complex to the inactive X chromosome in differentiating XX ES cells and PRC2 recruitment to target genes in undifferentiated ES cells. Required to repress Hox genes by enhancing H3K27me3 methylation of the PRC2 complex. In some conditions may act as an inhibitor of PRC2 activity: able to activate the CDKN2A gene and promote cellular senescence by suppressing the catalytic activity of the PRC2 complex locally. Binds to the metal-regulating-element (MRE) of MT1A gene promoter.
Especificaciones
| MTF2 | |
| Polyclonal | |
| Unconjugated | |
| Mtf2 | |
| 9230112N11Rik; AA537621; C76717; dJ976O13.2; hPCl2; M96; Metal regulatory transcription factor 2; metal response element binding transcription factor 2; metal response element DNA-binding protein M96; metal response element-binding transcription factor 2; metal-response element DNA-binding protein M96; Metal-response element-binding transcription factor 2; mPCl2; MTF2; PCL2; polycomb-like 2; polycomb-like protein 2; putative DNA binding protein; RGD1304727; RP5-976O13.1; TDRD19A; tudor domain containing 19A; zinc-regulated factor 1; ZiRF1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17765, 22823, 360905 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02395, Q9Y483 | |
| Mtf2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human MTF2 (NP_0011578641). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.