missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-56629
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MTF2 Polyclonal specifically detects MTF2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Especificaciones
| MTF2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| metal response element binding transcription factor 2, PCL2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| MTF2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GRTEGTAHSSNTSDVDFTGASSAKETTSSSISRHYGLSDSRKRTRTGRSWPAAIPHLRRRRGRLPRRALQTQNSEIVKDDE | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 22823 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido