missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTCP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA549474
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MTCP1 Polyclonal Antibody for Western Blot
This gene was identified by involvement in some t(X;14) translocations associated with mature T-cell proliferations. This region has a complex gene structure, with a common promoter and 5' exon spliced to two different sets of 3' exons that encode two different proteins. This gene represents the upstream 13 kDa protein that is a member of the TCL1 family. This protein may be involved in leukemogenesis.
Especificaciones
| MTCP1 | |
| Polyclonal | |
| Unconjugated | |
| MTCP1 | |
| C6.1b; C61B; Cmc4; mature T cell proliferation 1; mature T-cell proliferation 1; mature T-cell proliferation 1 isoform p13; mature T-cell proliferation 1 neighbor protein; mature T-cell proliferation-1 type A; mature T-cell proliferation-1 type B1; MTCP1; MTCP-1 type A; MTCP-1 type B1; p13MTCP1; p8MTCP1; Protein p13 MTCP-1; protein p8 MTCP-1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 17763, 4515 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P56278, Q60945 | |
| MTCP1 | |
| KLH conjugated synthetic peptide between 73-103 amino acids from human MTCP1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido