Learn More
MTCH2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16816654 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16816654 20 μL 20 microlitros
16806654 150 μL 150 microlitros
2 options
Código de producto. 16816654
Marca: Proteintech 168881AP20UL
Rabbit Polyclonal Antibody
MTCH2 is a member of the mitochondrial carrier protein family that catalyze the exchange of substrates across the inner mitochondrial membrane and is targeted by Bid, a pro-apoptotic Bcl-2 family member, in response to apoptotic signals, suggesting that MTCH2 may play a key role in the mitochondrial apoptosis pathway. Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. The Bcl-2 family of proteins is comprised of critical regulators of apoptosis that can be divided into two classes: those that inhibit apoptosis and those that promote cell death.
Especificaciones
Antígeno MTCH2
Aplicaciones Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.24 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica MTCH2
N.º de referencia del gen Q791V5, Q9Y6C9
Alias de gen HSPC032, MIMP, MTCH2
Símbolos de los genes Mtch2
Especie del huésped Rabbit
Inmunógeno MTCH2 Fusion Protein Ag10399
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23788, 56428
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG