Learn More
MTCH2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16888-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MTCH2 is a member of the mitochondrial carrier protein family that catalyze the exchange of substrates across the inner mitochondrial membrane and is targeted by Bid, a pro-apoptotic Bcl-2 family member, in response to apoptotic signals, suggesting that MTCH2 may play a key role in the mitochondrial apoptosis pathway. Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. The Bcl-2 family of proteins is comprised of critical regulators of apoptosis that can be divided into two classes: those that inhibit apoptosis and those that promote cell death.Especificaciones
| MTCH2 | |
| Polyclonal | |
| Unconjugated | |
| MTCH2 | |
| HSPC032, MIMP, MTCH2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23788, 56428 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q791V5, Q9Y6C9 | |
| Mtch2 | |
| MTCH2 Fusion Protein Ag10399 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |