missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MT-ND3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15741872 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15741872 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15741872 Proveedor Invitrogen™ N.º de proveedor PA564104

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: GYIEKSTPYE CGFDPISPAR VPFSIK Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.

NADH:ubiquinone oxidoreductase (complex I) is an extremely complicated multiprotein complex located in the inner mitochondrial membrane. Human complex I is important for energy metabolism because its main function is to transport electrons from NADH to ubiquinone, which is accompanied by trans-location of protons from the mitochondrial matrix to the intermembrane space. Human complex I appears to consist of 41 subunits. A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 3 (ND3) localizes to the hydrophobic protein fragment of complex I. Mutations in the gene encodiing for ND3 may be associated with Parkinson disease.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MT-ND3
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Polyclonal
Concentración 0.05 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica MT-ND3
N.º de referencia del gen P03897
Alias de gen 1.6.5.3; mitochondrially encoded NADH dehydrogenase 3; Mtnd3; mt-Nd3; NADH dehydrogenase subunit 3; NADH3; NADH-ubiquinone oxidoreductase chain 3; ND3
Símbolos de los genes MT-ND3
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human MT-ND3. Recombinant protein control fragment (Product #RP-103501).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4537
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.