Learn More
Invitrogen™ MSX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ 720172
Descripción
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Horse and Rabbit.
MSX1 is a member of the muscle segment homeobox gene family. It functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. MSX1 may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in the gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia.
Especificaciones
| MSX1 | |
| Polyclonal | |
| Unconjugated | |
| MSX1 | |
| AA675338; AI324650; ECTD3; homeo box, msh-like 1; homeobox 7; homeobox protein Hox-7; Homeobox protein MSX-1; homeobox, msh-like 1; Hox7; Hox-7; Hox7.1; hox-7.1; HYD1; msh; msh homeo box 1; msh homeobox 1; msh homeobox 1-like protein; msh homeobox homolog 1; Msx1; muscle-segment homeobox; STHAG1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
| P28360 | |
| MSX1 | |
| Protein corresponding to Human MSX1 (aa 1-303). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.