missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MST3 Monoclonal Antibody (ZM003)

Descripción
MST3 Monoclonal Antibody for Western Blot, ICC/IF, ELISA
STK24 (MST3) is a serine/threonine kinase that is a member of the GCK subfamily of STE20-like proteins. The yeast 'Sterile 20' gene (STE20) functions upstream of the mitogen-activated protein kinase (MAPK) cascade. In mammals, protein kinases related to STE20 can be divided into 2 subfamilies based on their structure and regulation. Members of the PAK subfamily (see PAK3; MIM 300142) contain a C-terminal catalytic domain and an N-terminal regulatory domain that has a CDC42 (MIM 116952)-binding domain. In contrast, members of the GCK subfamily (see MAP4K2; MIM 603166), also called the Sps1 subfamily, have an N-terminal catalytic domain and a C-terminal regulatory domain without a CDC42-binding domain. STK24 belongs to the GCK subfamily of STE20-like kinases.
Especificaciones
Especificaciones
| Antígeno | MST3 |
| Aplicaciones | ELISA, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | ZM003 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide; pH 7.4 |
| génica | STK24 |
| N.º de referencia del gen | Q9Y6E0 |
| Alias de gen | 1810013H02Rik; C76483; epididymis secretory protein Li 95; HEL-S-95; Mammalian STE20-like protein kinase 3; Mammalian STE20-like protein kinase 3 C-terminal; Mammalian STE20-like protein kinase 3 N-terminal; mammalian sterile twenty 3 kinase; Mst3; MST-3; MST3/C; MST3/N; MST3B; OTTHUMP00000018594; RGD1561742; RP11-111L24.5; serine/threonine kinase 24; serine/threonine kinase 24 (STE20 homolog, yeast); serine/threonine-protein kinase 24; Serine/threonine-protein kinase 24 12 kDa subunit; Serine/threonine-protein kinase 24 35 kDa subunit; Serine/threonine-protein kinase 24 36 kDa subunit; STE20; STE20-like kinase 3; STE20-like kinase MST3; sterile 20-like kinase 3; STK24; STK3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?