Learn More
MSP/MST1 Antibody (S04-1D6), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00€ - 645.00€
Especificaciones
Antígeno MSP/MST1
Clon S04-1D6
Concentración 0.3 mg/mL
Dilución Western Blot, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
18655903
Ver documentos
Novus Biologicals
NBP3-19812-100ul
100 μg
645.00€
100 microlitros
Ver productos alternativos
Ahorre hasta 
»
18686043
Ver documentos
Novus Biologicals
NBP3-19812-50ul
50 μg
421.00€
50 microlitros
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
MSP/MST1 Monoclonal antibody specifically detects MSP/MST1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
MSP/MST1
0.3 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Protein Kinase
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
4485
IgG
Affinity purified
S04-1D6
Western Blot, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, NF15S2
A synthetic peptide of human MSP/MST1 (Uniprot # P26927)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.