missing translation for 'onlineSavingsMsg'
Learn More

MRS-1706, MedChemExpress

Código de producto. 16476527
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
Código de producto. Cantidad unitSize
16476527 5 mg 5 miligramos
16444856 100 mg 100 miligramos
16413808 10 mg 10 miligramos
16400187 50 mg 50 miligramos
4 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 16476527

missing translation for 'mfr': Medchem Express HY103186

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress MRS-1706 is a potent and selective adenosine A2B receptor inverse agonist. MRS-1706 has Ki values of 1.39, 112, 157, and 230 nM for human A2B, A2A, A1 and A3 receptors respectively. MRS-1706 blocks adenosine-mediated cAMP induction.

MRS-1706 is a potent and selective adenosine A2B receptor inverse agonist. MRS-1706 has Ki values of 1.39, 112, 157, and 230 nM for human A2B, A2A, A1 and A3 receptors respectively. MRS-1706 blocks adenosine-mediated cAMP induction.

Specifications

Nombre del producto químico o material MRS-1706
CAS 264622-53-9
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C27H29N5O5
Cantidad 5 mg
Información de solubilidad DMSO : 240 mg/mL (476.62 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C(C)=O)C=C1)COC2=CC=C(C3=NC(N(CCC)C(N(CCC)C4=O)=O)=C4N3)C=C2
Peso molecular (g/mol) 503.55
Formula Weight (peso de la fórmula) 503.55
Porcentaje de pureza 98.23%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.