missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS22 Polyclonal Antibody

Descripción
The antibody detects endogenous levels of total MRPS22 protein.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that does not seem to have a counterpart in prokaryotic and fungal-mitochondrial ribosomes. This gene lies telomeric of and is transcribed in the opposite direction from the forkhead box L2 gene. A pseudogene corresponding to this gene is found on chromosome Xq.
Especificaciones
Especificaciones
| Antígeno | MRPS22 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | MRPS22 |
| N.º de referencia del gen | P82650 |
| Alias de gen | 28S ribosomal protein S22, mitochondrial; 3100002P07Rik; C3orf5; COXPD5; GIBT; GK002; mitochondrial ribosomal protein S22; Mitochondrial small ribosomal subunit protein mS22; MRPS22; MRP-S22; ribosomal protein, mitochondrial, S22; Rpms22; S22mt |
| Símbolos de los genes | MRPS22 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?