Learn More
MRPL45 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16834964 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16834964 20 μL 20 microlitros
16824964 150 μL 150 microlitros
2 options
Código de producto. 16834964
Marca: Proteintech 156821AP20UL
Rabbit Polyclonal Antibody
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Pseudogenes corresponding to this gene are found on chromosomes 2p and 17q.
Especificaciones
Antígeno MRPL45
Aplicaciones Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation
Clasificación Polyclonal
Concentración 0.16 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica MRPL45
N.º de referencia del gen Q9BRJ2, Q9D0Q7
Alias de gen L45mt, MRPL45, MRP-L45
Símbolos de los genes Mrpl45
Especie del huésped Rabbit
Inmunógeno MRPL45 Fusion Protein Ag8184
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 287656, 67036, 84311
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG