missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL12 Polyclonal Antibody

Descripción
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.
MRPL12 (mitochondrial ribosomal protein L12), also referred to as 5c5-2, L12mt, MRPL7 or RPML12, is a mammalian mitochondrial ribosomal protein that is involved in protein synthesis within the mitochondrion. MRPL12 is enhanced in growth-stimulated cells as a result of transcriptional activation, suggesting that it may function as a translational regulator of mitochondrial mRNAs. Impairment of MRPL12 leads to reduction in cell growth rate, decreased mitochondrial ATP production and abolition of mitochondrial oxidative phosphorylation. MRPL12 is cleaved during its translocation across the mitochondrial membrane and it exists as dimers that bind the large ribosomal subunit. MRPL12 is 198 amino acids in length, belongs to the ribosomal protein L12P family and is highly expressed in the colon.
Especificaciones
Especificaciones
| Antígeno | MRPL12 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | MRPL12 |
| N.º de referencia del gen | 0 |
| Alias de gen | 0610034O11Rik; 1500031N16Rik; 39S ribosomal protein L12, mitochondrial; 5c5-2; FLJ60124; hypothetical protein LOC550391; im:6893415; L12mt; MGC8610; Mitochondrial large ribosomal subunit protein bL12m; mitochondrial ribosomal protein L12; mitochondrial ribosomal protein L7/12; MRPL12; MRP-L12; MRP-L31/34; MRPL7; MRPL7/L12; ribosomal protein, mitochondrial, L12; Rpm12; RPML12; zgc:112445 |
| Símbolos de los genes | MRPL12 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?