Learn More
Invitrogen™ MRP3 Monoclonal Antibody (M3II-21)

Descripción
The antibody reacts with an internal epitope of MRP3, a 190-200 kD transmembrane protein that is closely related to the multidrug resistance protein MRP1. M3II-21 was raised against a bacterial fusion protein of MRP3, containing amino acids 830-949 of the protein. M3II-21 does not cross-react with the human MDR1, MRP1, MRP2 or MRP5 gene products. M3II-21 has potential value for detection of MRP3-mediated drug-resistance in human tumor samples.
Especificaciones
Especificaciones
| Antígeno | MRP3 |
| Aplicaciones | Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | M3II-21 |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | Tissue culture supernatant with 0.7% BSA and 0.1% sodium azide |
| génica | ABCC3 |
| N.º de referencia del gen | O15438 |
| Alias de gen | ABC31; ABCC3; ATP binding cassette subfamily C member 3; ATP-binding cassette sub-family C member 3; ATP-binding cassette, subfamily C (CFTR/MRP), member 3; ATP-binding cassette, sub-family C (CFTR/MRP), member 3; canalicular multispecific organic anion transporter 2; canicular multispecific organic anion transporter; cMOAT2; DKFZp686E22157; EST90757; MLP2; MLP-2; MOAT-D; Mrp3; MRP-like protein 2; multidrug resistance associated protein; multidrug resistance protein 3; multidrug resistance-associated protein 3; multi-specific organic anion transporter D; organic anion transporter |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.