missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRP2 Monoclonal Antibody (M2III-5)
GREENER_CHOICE

Código de producto. 11564982
Change view
Click to view available options
Cantidad:
1 mL
Tamaño de la unidad:
1 ml
Código de producto. Cantidad unitSize
11564982 1 mL 1 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11564982

Marca: Invitrogen™ MA135678

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The antibody reacts with an internal epitope of MRP2, a 190-200 kD transmembrane protein earlier known as the canalicular multi-organic anion transporter cMOAT, absent in patients with the Dubin-Johnson syndrome, an autosomal recessive liver disorder characterized by chronic conjugated hyperbilirubinemia. MRP2 is a member of the MRP family of multidrug resistance related proteins, and MRP2 overexpression has been observed in a subset of cisplatin resistant cell lines. M2III-5 was raised against a fusion protein of the bacterial maltose binding protein and rat Mrp2, containing the 202-amino acid COOH terminal end of the transporter protein. The Mab detects rat, mouse and human MRP2. M2III-5 does not cross-react with the human MDR1 P-gp, MRP1, MRP3 orMRP5 gene products.

This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MRP2
Aplicaciones Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon M2III-5
Concentración Conc. Not Determined
Conjugado Unconjugated
Formulación Tissue culture supernatant with 0.7% BSA and 0.1% sodium azide
génica ABCC2
N.º de referencia del gen Q63120, Q8VI47, Q92887
Alias de gen ABC30; Abcc2; AI173996; ATP binding cassette subfamily C member 2; ATP-binding cassette sub-family C member 2; ATP-binding cassette, subfamily C (CFTR/MRP), member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; calicular multispecific organic anion transporter; canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; Cmoat; CMOAT1; Cmrp; DJS; Human MRP2; KIAA1010; MRP2; multidrug resistance associated protein 2; multidrug resistance protein 2; multidrug resistance-associated protein 2
Símbolos de los genes ABCC2
Especie del huésped Mouse
Inmunógeno 202-amino acid COOH terminal of rat Mrp2.
Cantidad 1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1244, 12780, 25303
Especies diana Human, Mouse, Rat
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.