missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRG Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15751471 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15751471 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15751471 Proveedor Invitrogen™ N.º de proveedor PA553534

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: LFHAMLGHKP VGVNRHFHMI CIRDKFSQNI GRQVPSKVIW DHLSTMYDMQ ALHESEILPF PNPERNFVLP EEIIQEVREG KVMIEEEMKE EMKEDVDPHN GADDVFSSSG SLGKASEKSS KDKEKNSSDL GCKE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

C20orf20, also named as MRG/MORF4L-binding protein or Up-regulated in colon cancer 4 (Urcc4), is a 204 amino acid protein, which belongs to the EAF7 family. C20orf20 localizes in the nucleus and is a Component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. C20orf20 is modified through phosphorylation and acetylation, so the molecular weight of C20orf20 might be 25-30 kDa.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MRG
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.4 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica MRGBP
N.º de referencia del gen Q9DAT2, Q9NV56
Alias de gen 1600027N09Rik; AW060503; C20orf20; C80444; Eaf7; FLJ10914; MRG/MORF4L binding protein; MRG/MORF4L-binding protein; MRG15BP; MRG-binding protein; MRGBP; RGD1308612; Up-regulated in colon cancer 4; Urcc4
Símbolos de los genes MRGBP
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human MRG. Recombinant protein control fragment (Product #RP-89545).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 311713, 55257, 73247
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.