Learn More
MRE11A Mouse anti-Human, Unconjugated, Clone: 1B12B2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67570-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The MRE11A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MRE11A. This antibody recognizes human, mouse, rat antigen. The MRE11A antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| MRE11A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 1B12B2 | |
| Unconjugated | |
| 150 μL | |
| 4361 | |
| IgG1 |