Learn More
CPC (Colder) MPX™ Quick-Disconnect Fittings, Plug
Click to view available options
Color:
Tinción ámbar
Tinte púrpura
Tamaño de la unidad:
paquete de 25
Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Temperatura mín. de funcionamiento (métrico) | -40°F |
| Puerto de salida | Plug |
| Método de esterilización | 121°C |
| Forma | Plug |
| Material (sello) | Silicone |
| Color | Tinte púrpura |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.