Learn More
CPC (Colder) MPX™ Quick-Disconnect Fittings, Hose Barb Body, Non-Valved
Descripción
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Connect to a male coupling inserts for easy media transfer. Operate ergonomic thumb latch even with gloved hands. Mix and match termination sizes for added flexibility. 0.5 in. nominal flow path size
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diámetro interior (imperial) | 1/2 in. |
| Material | Policarbonato |
| Temperatura mín. de funcionamiento (métrico) | -40°F |
| Cantidad de embalaje | 25/Pk. |
| Manguera dentada | Cuerpo de manguera dentada |
| Puerto de salida | Quick Disconnect Body x Hose Barb |
| Método de esterilización | 121°C |
| Forma | Straight |
| Color | Tinte púrpura |