Learn More
MPV17L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17884-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
17884-1-AP is a rabbit polyclonal antibody raised against the full-length of human MPV17L.
Isoform 1 participates in reactive oxygen species metabolism by up- or down-regulation of the genes of antioxidant enzymes.Especificaciones
| MPV17L | |
| Polyclonal | |
| Unconjugated | |
| MPV17L | |
| M LP homolog, M LPH, MLPH1, MLPH2, Mpv17 like protein, MPV17L, MPV17L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 255027, 93734 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q2QL34, Q99MS3 | |
| Mpv17l | |
| MPV17L Fusion Protein Ag11935 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |