Learn More
Invitrogen™ MPST Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA588156
Descripción
Immunogen sequence: LVSAQWVAEA LRAPRAGQPL QLLDASWYLP KLGRDARREF EERHIPGAAF FDIDQCSDRT SPYDHMLPGA EHFAEYAGRL GVGAATHVVI YDASDQGLYS APRVWWMFRA FGHHAVSLLD GGLRHWLRQN LPLSSGKSQP A; Positive Samples: Mouse liver, Rat liver; Cellular Location: Cell junction, Cytoplasm, Mitochondrion, synapse, synaptosome.
This protein encoded by this gene catalyzes the transfer of a sulfur ion from 3-mercaptopyruvate to cyanide or other thiol compounds. It may be involved in cysteine degradation and cyanide detoxification. There is confusion in literature between this protein (mercaptopyruvate sulfurtransferase, MPST), which appears to be cytoplasmic, and thiosulfate sulfurtransferase (rhodanese, TST, GeneID:7263), which is a mitochondrial protein. Deficiency in MPST activity has been implicated in a rare inheritable disorder known as mercaptolactate-cysteine disulfiduria (MCDU). Alternatively spliced transcript variants encoding same or different isoforms have been identified for this gene.
Especificaciones
| MPST | |
| Polyclonal | |
| Unconjugated | |
| Mpst | |
| 3-mercaptopyruvate sulfurtransferase; human liver rhodanese; mercaptopyruvate sulfurtransferase; MPST; MST; testicular tissue protein Li 200; tRNA thiouridin modification protein 1; TST2; TUM1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192172, 246221 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.28 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P97532, Q99J99 | |
| Mpst | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-170 of human MPST (NP_0669492). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.