missing translation for 'onlineSavingsMsg'
Learn More

MPP3 Monoclonal Antibody (OTI2A11), TrueMAB™, OriGene

Código de producto. p-200009805
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17514591 30 μL 30 microlitros
17594581 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17514591 Supplier Origene Technologies Supplier No. TA501564S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MPP3 Monoclonal antibody specifically detects MPP3 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antígeno MPP3
Aplicaciones Immunocytochemistry, Western Blot
Clasificación Monoclonal
Clon OTI2A11
Concentración 0.54 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica Prpsap2
N.º de referencia del gen O60256
Alias de gen 6430514B01; CSG18; discs large homolog 3; discs, large homolog 3; DLG3; Dlgh3; Dusp3; MAGUK p55 subfamily member 3; MAGUK p55 subfamily member 3 {ECO:0000250; membrane palmitoylated protein 3; membrane protein palmitoylated 3; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3); Mpp3; mpp3 {ECO:0000312; Mpp3 membrane protein, palmitoylated 3; protein Dlgh3; protein MPP3; RGD:620015}; UniProtKB:O88910}
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human PRPSAP2 produced in HEK293T cell.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5636
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.