Learn More
R&D Systems™ Mouse TNF-alpha Quantikine ELISA Kit
Código de producto. 30226445
Click to view available options
Cantidad:
1 placa
2 placas
6 placas
Tamaño de la unidad:
1 set
Código de producto. 30226445
Marca: R&D Systems™ MTA00B1
Mouse TNF-alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Mouse TNF-alpha Quantikine ELISA Kit quantifies mouse TNF-alpha in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY
Especificaciones
Intervalo del ensayo 10.90 - 700pg/mL
Sensibilidad del ensayo 7.21pg/mL
Conjugado HRP
Tipo de producto Solid Phase Sandwich ELISA
Tipo de muestra Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Especificidad Natural and Recombinant Mouse TNF-alpha
Suficiente para 2 x 96-well Plates
Especies diana Mouse
Para utilizar con (aplicación) ELISA
ID de gen (Entrez) 21926
Cantidad 1 placa
Volumen de muestra Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Requisitos de almacenamiento Store the unopened product at 2° to 8°C. Do not use past expiration date.
Sinónimo APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha