Learn More
Mouse TLR3 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18111661
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18111661 100 μg 100 microgramos
18719038 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18111661 Proveedor R&D Systems N.º de proveedor MAB3005
Rat Monoclonal Antibody has been used in 2 publications
TLR3 Monoclonal specifically detects TLR3 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TLR3
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 313129
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen Q99MB1
Alias de gen CD283, CD283 antigen, toll-like receptor 3
Símbolos de los genes TLR3
Especie del huésped Rat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant mouse TLR3 Cys29-Leu705 Accession # Q99MB1
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7098
Especificidad de la prueba Detects mouse TLR3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TLR1, 2, 4, 6, recombinant human TLR3 or 4 is observed.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2a
Mostrar más Mostrar menos
For Research Use Only