missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TCF-3/E2A Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems MAB7650-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TCF-3/E2A Monoclonal specifically detects TCF-3/E2A in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
| TCF-3/E2A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P15806 | |
| TCF3 | |
| E. coli-derived recombinant mouse TCF-3/E2A Asn33-Arg159 Accession # P15806 | |
| 25 μg | |
| Primary | |
| Detects mouse TCF-3/E2A in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human TCF-3 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 826927 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BHLHB21, bHLHb21TCF-3, Class B basic helix-loop-helix protein 21, E2A, E2A immunoglobulin enhancer-binding factor E12/E47, E2AKappa-E2-binding factor, Immunoglobulin enhancer-binding factor E12/E47, Immunoglobulin transcription factor 1, ITF1, ITF1Transcription factor ITF-1, Kappa-E2-binding factor, MGC129647, MGC129648, Pan2, Tcfe2a, Transcription factor 3, transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47), transcription factor E2-alpha, VDIR, VDR interacting repressor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6929 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido