missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-E Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems FAB5806T-100UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Siglec-E Monoclonal specifically detects Siglec-E in Mouse samples. It is validated for Flow Cytometry.
Especificaciones
| Siglec-E | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q91Y57 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Siglec-E in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Siglec-3/CD33 or rmSiglec-F is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 750620 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Siglece | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-E Gln19-Phe354 Accession # Q91Y57 | |
| RUO | |
| 8778 | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction