missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Siglec-1/CD169 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marca: R&D Systems AF5610
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Siglec-1/CD169 Polyclonal specifically detects Siglec-1/CD169 in Mouse samples. It is validated for Western Blot, Neutralization.
Especificaciones
| Siglec-1/CD169 | |
| Polyclonal | |
| Unconjugated | |
| Q62230 | |
| SIGLEC1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-1/CD169 Thr20-Leu1639 Accession # Q62230 | |
| 100 μg | |
| Primary | |
| Detects mouse Siglec-1/CD169 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) Siglec-1 is observed and less than 1% cross-reactivity with rhSiglec-2 and recombinant mouse Siglec-2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 1-5 ug/mL | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin, Siglec1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6614 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido