missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Serpin E1/PAI-1 DuoSet ELISA

Código de producto. 30001713
Change view
Click to view available options
Cantidad:
5 placas
Tamaño de la unidad:
paquete de 5
Código de producto. Cantidad unitSize
30001713 5 placas paquete de 5
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 30001713

Brand: R&D Systems™ DY382805

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Serpin E1/PAI-1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Serpin E1/PAI-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Serpin E1/PAI-1.
TRUSTED_SUSTAINABILITY

Specifications

Analitos detectables Serpin E1/PAI-1
ensayo ELISA Development Kit
Conjugado Biotina
Reactividad cruzada < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Formato del kit de inmunoensayo 96-well microplate (sold separately)
Para utilizar con (aplicación) ELISA
ID de gen (Entrez) 18787
Especie del huésped Ratón
Intervalo de medición 9.38-600pg/mL
Cantidad 5 placas
Requisitos de almacenamiento Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.