Learn More
Mouse S100A4 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18733989
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
18733989 25 μg 25 microgramos
1 options
Código de producto. 18733989 Proveedor R&D Systems N.º de proveedor MAB4138SP
Rat Monoclonal Antibody
S100A4 Monoclonal specifically detects S100A4 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno S100A4
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 438709
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 2 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P07091
Alias de gen 18A2, 42A, Calvasculin, CAPL, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog)
Símbolos de los genes S100A4
Especie del huésped Rat
Inmunógeno E. coli-derived recombinant mouse S100A4 Ala2-Lys101 Accession # P07091
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6275
Especificidad de la prueba Detects mouse S100A4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse S100A6, 8, 9, 10, 13, 16, recombinant human (rh) S100A1, 2, 7, 11, or rhS100P is observed.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2b
Mostrar más Mostrar menos
For Research Use Only