missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PLUNC Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Marca: R&D Systems AF4274
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PLUNC Polyclonal specifically detects PLUNC in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
| PLUNC | |
| Polyclonal | |
| Unconjugated | |
| NP_035256 | |
| BPIFA1 | |
| E. coli-derived recombinant mouse PLUNC Ala59-Val278 Accession # NP_035256 | |
| 100 μg | |
| Primary | |
| Detects mouse PLUNC in direct ELISAs and Western blots. In these formats, less than 10% cross-reactivity with recombinant human PLUNC is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| bA49G10.5, BPIFA1, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNX, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 51297 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?