Learn More
Mouse PD-L1/B7-H1 Antibody, R&D Systems™
Código de producto. 30306072
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
30306072 25 μg 25 microgramos
30306087 100 μg 100 microgramos
2 options
Código de producto. 30306072
Marca: R&D Systems MAB11630SP
Rabbit Monoclonal Antibody
PD-L1 Monoclonal antibody specifically detects PD-L1 in Mouse samples. It is validated for Immunohistochemistry,Multiplex Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PD-L1
Aplicaciones Immunohistochemistry, Multiplex Immunofluorescence
Clasificación Monoclonal
Clon 2096A
Conjugado Unconjugated
Dilución Immunohistochemistry 0.05-10 μg/mL, Multiplex Immunofluorescence 5 μg/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
N.º de referencia del gen Q9EP73
Alias de gen Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Especie del huésped Rabbit
Inmunógeno Mouse myeloma cell line, NS0 derived mouse PD-L1/B7-H1, Phe19-His239, Accession # Q9EP73
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 29126
Reconstitución Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.