missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin alpha 7 PE-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 4 publications
Marca: R&D Systems FAB3518P
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Integrin alpha 7 Monoclonal specifically detects Integrin alpha 7 in Mouse samples. It is validated for Flow Cytometry.
Especificaciones
| Integrin alpha 7 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FLJ25220, integrin alpha 7 chain, integrin alpha-7, integrin, alpha 7, ITGA7 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3679 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 334908 | |
| Flow Cytometry 10 uL/10^6 cells | |
| NP_032424 | |
| ITGA7 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 7 Phe34-Pro1036 Accession # NP_032424 | |
| 100 Tests | |
| Primary | |
| Detects mouse Integrin alpha 7 in direct ELISAs. Reactivity with other Integrin alpha 7 isoforms was not tested. In direct ELISAs, no cross-reactivity with recombinant mouse Integrin alpha 2, alpha 3, alpha 4, alpha 5, alpha 9, alpha E, alpha X, recombinant human Integrin alpha L, or alpha 11 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido