missing translation for 'onlineSavingsMsg'
Learn More

Mouse Integrin alpha 10 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18793589
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
18793589 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18793589

Marca: R&D Systems FAB9757V100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

Integrin alpha 10 Monoclonal specifically detects Integrin alpha 10 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Integrin alpha 10
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 885501
Conjugado Alexa Fluor 405
Dilución Flow Cytometry 0.25-1 ug/10^6 cells
Formulación Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
N.º de referencia del gen NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
Alias de gen integrin alpha-10, integrin, alpha 10, ITGA10, PRO827
Especie del huésped Rat
Inmunógeno Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1 Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1) Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Primario o secundario Primary
ID de gen (Entrez) 8515
Especificidad de la prueba Detects mouse Integrin alpha 10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Intagrin alpha 2 beta 1 was observered.
Especies diana Mouse
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Formulario Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.