Learn More
Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™
Código de producto. 18740047
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18740047 100 μg 100 microgramos
18711717 25 μg 25 microgramos
2 options
Código de producto. 18740047
Marca: R&D Systems MAB22972100
Rat Monoclonal Antibody
IL-36 alpha/IL-1F6 Monoclonal antibody specifically detects IL-36 alpha/IL-1F6 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IL-36 alpha/IL-1F6
Aplicaciones ELISA
Clasificación Monoclonal
Clon 275302
Conjugado Unconjugated
Dilución ELISA
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen Q9JLA2
Alias de gen FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha
Especie del huésped Rat
Inmunógeno E. coli-derived recombiMouset mouse IL-36 alpha/IL-1F6, Met1-His160, Accession # Q9JLA2
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 27179
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2a