missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-12 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18746238
Change view
Click to view available options
Cantidad:
25 μg
Unit Size:
25 microgramos
Product Code. Cantidad unitSize
18746238 25 μg 25 microgramos
1 options
This item is not returnable. View return policy

Product Code. 18746238

Brand: R&D Systems AF419SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 7 publications

IL-12 Polyclonal specifically detects IL-12 in Mouse samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IL-12
Aplicaciones Western Blot, Neutralization
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Neutralization 0.01-0.06 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Alias de gen CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Símbolos de los genes IL12A
Especie del huésped Goat
Inmunógeno S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-12
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3592
Especificidad de la prueba Detects mouse IL-12 in direct ELISAs and Western blots. In direct ELISAs and Western blots (non-reducing and reducing conditions), this antibody shows less than 10% cross-reactivity with recombinant human IL-12.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.