Learn More
Mouse IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18765199
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18765199 25 μg 25 microgramos
18798898 100 μg 100 microgramos
2 options
Código de producto. 18765199
Marca: R&D Systems MAB8234SP
Rabbit Monoclonal Antibody
IFN-beta Monoclonal specifically detects IFN-beta in Mouse samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IFN-beta
Aplicaciones Neutralization
Clasificación Monoclonal
Clon 1176D
Conjugado Unconjugated
Dilución Neutralization 0.25-1.25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen P01575
Alias de gen Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Especie del huésped Rabbit
Inmunógeno Human embryonic kidney cell line HEK293-derived mouse IFN-beta Ile22-Asn182 Accession # P01575
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3456
Especificidad de la prueba Detects mouse IFN-beta in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG