missing translation for 'onlineSavingsMsg'
Learn More

Mouse IFN-alpha/beta R2 Fluorescein-conjugated Antibody, R&D Systems™

Product Code. 18125611
Change view
Click to view available options
Cantidad:
100 Tests
Unit Size:
100 tests
Product Code. Cantidad unitSize
18125611 100 Tests 100 tests
1 options
This item is not returnable. View return policy

Product Code. 18125611

Brand: R&D Systems FAB1083F

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody

IFN-alpha/beta R2 Polyclonal specifically detects IFN-alpha/beta R2 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IFN-alpha/beta R2
Aplicaciones Flow Cytometry
Clasificación Polyclonal
Conjugado Fluorescein
Dilución Flow Cytometry 10 uL/10^6 cells
N.º de referencia del gen Q9D1R7
Alias de gen beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFN-aR2, IFNAR2, IFNARB, IFNbR2, IFN-bR2, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2
Símbolos de los genes IFNAR2
Especie del huésped Goat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant mouse IFN-alpha/beta R2 Ser22-Ala239 (Lys160Asn) Accession # Q9D1R7
Método de purificación Affinity Purified
Cantidad 100 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3455
Especificidad de la prueba Detects mouse IFN-alpha/beta R2 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) IFN-gamma R2 and rmIL-20 R alpha is observed, 5% cross-reactivity with recombinant human (rh) IL-10 R beta , rmIFN-gamma RI, and rhIL-20 R beta is observed and less than 1% cross-reactivity with rhIFN-beta R alpha is observed.
Especies diana Mouse
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.