missing translation for 'onlineSavingsMsg'
Learn More

Mouse Gr-1/Ly-6G PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18795238
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Unit Size:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18795238 25 Tests 25 tests
18726936 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18795238 Supplier R&D Systems Supplier No. FAB1037C025

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Gr-1/Ly-6G
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon RB6-8C5
Conjugado PerCP
Dilución Flow Cytometry 10 uL/10^6 cells
Formulación Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Alias de gen C6orf23, G6D, Ly-6C, LY6-D, Ly6G, Ly-6G, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Símbolos de los genes Ly6g
Especie del huésped Rat
Inmunógeno The immunogen for this antibody is Ly-6G (Gr-1, Gr1).
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 58530
Especificidad de la prueba Detects mouse Gr-1/Ly-6G. Weak cross-reactivity with Ly-6C is observed.
Especies diana Mouse
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.