missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems BAF639
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
EphA2 Polyclonal specifically detects EphA2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Spécification
| EphA2 | |
| Polyclonal | |
| Biotin | |
| AAA82113 | |
| EPHA2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA2 Ala22-Ala535 Accession # AAA82113 | |
| 50 μg | |
| Primary | |
| Detects mouse EphA2 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human EphA1, recombinant mouse (rm) EphA3, rmEphA4, rmEphA6, rmEphA7, rmEphA8 and recombinant rat EphA5 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| ARCC2, EC 2.7.10, EC 2.7.10.1, Eck, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, Myk2, Sek2, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1969 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu