missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse E-Selectin/CD62E Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Marca: R&D Systems MAB5751-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Especificaciones
| E-Selectin/CD62E | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6401 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, ELISA | |
| 96419 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q00690 | |
| SELE | |
| Mouse myeloma cell line NS0-derived recombinant mouse E-Selectin Trp22-Pro557 Accession # Q00690 | |
| 25 μg | |
| Primary | |
| Detects mouse E-Selectin in ELISAs and Western blots. Does not cross-react with recombinant human (rh) E-Selectin, rmL-Selectin, or rhP-Selectin. | |
| Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido