missing translation for 'onlineSavingsMsg'
Learn More

Mouse CX3CR1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18708118
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18708118 25 Tests 25 tests
18775857 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Codice prodotto. 18708118 Fornitore R&D Systems N. del fornitore FAB5825A025

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Goat Polyclonal Antibody has been used in 3 publications

CX3CR1 Polyclonal specifically detects CX3CR1 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifica

Antígeno CX3CR1
Aplicaciones Flow Cytometry
Clasificación Polyclonal
Conjugado APC
Dilución Flow Cytometry 10 uL/10^6 cells
N.º de referencia del gen Q9Z0D9
Alias de gen Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1
Símbolos de los genes CX3CR1
Especie del huésped Goat
Inmunógeno E. coli-derived recombinant mouse CX3CR1 Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274 Accession # Q9Z0D9
Método de purificación Affinity Purified
Cantidad 25 Tests
Primario o secundario Primary
ID de gen (Entrez) 1524
Especificidad de la prueba Detects mouse CX3CR1 in direct ELISAs. Stains mouse CX3CR1 transfectants but not irrelevant transfectants in flow cytometry.
Especies diana Mouse
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.