missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CTRP5/C1qTNF5 (NP_663588) VersaClone cDNA
Beskrivning
The Mouse CTRP5/C1qTNF5 (NP_663588) VersaClone cDNA clone contains the complete ORF for C1qTNF5/CTRP5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The C1qTNF5/CTRP5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifikationer
Specifikationer
| génica | mC1qtnf5 |
| Número de acceso | NP_663588 |
| Concentración | 10 μg at 0.2 μg/μL |
| Formulación | cDNA is provided in 10 mM Tris-Cl |
| Tipo de inserto | 745bp |
| pH | 8.5 |
| ID de gen (Entrez) | 114902 |
| Contenido y almacenamiento | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Especie | Mouse |
| Cantidad | 10 μg |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?