missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD83 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems AF1437
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD83 Polyclonal specifically detects CD83 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, Immunocytochemistry, CyTOF-ready.
Especificaciones
| CD83 | |
| Polyclonal | |
| Unconjugated | |
| O88324 | |
| CD83 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD83 Met22-Ala134 Accession # O88324 | |
| 100 μg | |
| Primary | |
| Detects mouse CD83 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Adhesion Blockade, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| B-cell activation protein, BL11, BL11CD83 antigen, CD83, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15, HB15hCD83 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9308 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido